Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Themis Monoclonal Antibody (1TMYS), PE, eBioscience™
GREENER_CHOICE

Catalog No. 12591880
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-591-880 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-591-880 Supplier Invitrogen™ Supplier No. 12591880
  • $193.00
    $144.65 / Each of 1

    Save $48.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

This 1TMYS monoclonal antibody reacts with mouse Thymocyte-expressed molecule involved in selection (Themis), also known as Grb2-associating protein (Gasp). Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing. The 1TMYS antibody recognizes isoforms 1, 2, 3, and 4, but does not recognize isoform 5. Applications Reported: This 1TMYS antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 1TMYS antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes or splenocytes using the intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cell...

Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Themis
Applications Flow Cytometry
Classification Monoclonal
Clone 1TMYS
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Themis
Gene Accession No. Q8BGW0
Gene Alias bA325O24.3; bA325O24.4; C6orf190; C6orf207; E430004N04Rik; FLJ40584; Gasp; GRB2-associated protein; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; RGD1560849; signaling phosphoprotein specific for T cells; SPOT; THEMIS; thylex; thymocyte expressed molecule involved in selection; thymocyte selection associated; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection; Tsepa
Gene Symbols Themis
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210757
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.