Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thenoyltrifluoroacetone, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
326-91-0
C8H5F3O2S
222.19
MFCD00005445
TXBBUSUXYMIVOS-UHFFFAOYSA-N
2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone
5601
4,4,4-trifluoro-1-thiophen-2-ylbutane-1,3-dione
C1=CSC(=C1)C(=O)CC(=O)C(F)(F)F
326-91-0
C8H5F3O2S
222.19
MFCD00005445
TXBBUSUXYMIVOS-UHFFFAOYSA-N
2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone
5601
4,4,4-trifluoro-1-thiophen-2-ylbutane-1,3-dione
C1=CSC(=C1)C(=O)CC(=O)C(F)(F)F
Acylation reagent
Boiling Point 96°C to 98°C (8.0mmHg)
Color Beige to Brown or Yellow
Flash Point 111°C
Quantity 25g
Packaging Glass bottle
Melting Point 40°C to 44°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble
Formula Weight 222.19
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity ≥98.5%
Chemical Name or Material Thenoyltrifluoroacetone
Show More Show Less

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning