Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC459251000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459251000 100 g
AC459255000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459251000 Supplier Thermo Scientific Chemicals Supplier No. 459251000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-37-9
Molecular Formula C9H9N4NaO4
Molecular Weight (g/mol) 260.19
MDL Number MFCD00022832
InChI Key MSFVZSOKOXZSME-UHFFFAOYSA-M
Synonym acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid
PubChem CID 69550
IUPAC Name 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid
SMILES [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O

Specifications

Melting Point 270.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Formula Weight 238.2
Percent Purity 98%
Chemical Name or Material Theophylline-7-acetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 211-490-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.