Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theophylline, Anhydrous, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18607341 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-341 100 g Amber Glass Bottle
18-607-342 500 g Amber Glass Bottle
18-607-343 2.5 kg Amber Glass Bottle
18-607-344 12 kg Amber Glass Bottle
18-607-345 25 kg Amber Glass Bottle
18-607-346 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-341 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TH110100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

TH110, 58-55-9, C7H8N4O2

Spectrum™ Chemical Theophylline, Anhydrous, USP is an oral medication used to treat asthma and other lung diseases. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-55-9
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.17
InChI Key ZFXYFBGIUFBOJW-UHFFFAOYSA-N
IUPAC Name 1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C2=C(NC=N2)C(=O)N(C)C1=O

Specifications

CAS Max % 1
Ignition Residue 0.0015
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Theophylline, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.