Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Aqui-Pad™ Benchtop Absorbent Mat

Catalog No. 22131401 Shop All Therapak Corporation Products
Change view
Click to view available options
Capacity (Metric):
750 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-131-401 750 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-131-401 Supplier Therapak™ Supplier No. 10324
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work.

  • Consists of two material layers with absorbent media sealed inside
  • Non-woven polypropylene pad top layer and impervious polypropylene film bottom layer to enclose any liquid spills so the liquid does not reach the benchtop
  • Inside the two layers is a proprietary absorbent media fabricated from tissue paper and wood pulp materials that are impregnated with a super absorbent polymer
  • Supplied in flat sheets 18 x 11 in. (457 x 280mm)
  • Absorbs up to 750mL
  • Placed in biohazard disposal after use

Specifications

Capacity (English) 0.19 gal.
Capacity (Metric) 750 mL
Material Polypropylene
Type Aqui-Pad™ Benchtop Absorbent Mat
For Use With (Application) For use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.