Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Biohazard Labels

Catalog No. 22130118 Shop All Therapak Corporation Products
Change view
Click to view available options
Size (Metric):
7.62 x 5.08 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
22-130-118 7.62 x 5.08 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-130-118 Supplier Therapak™ Supplier No. 54607
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Adhere to corrugated boxes and plastic containers

  • Warn employees and other handlers of the risk of infection from contact with biohazardous material
  • Kiss-cut on rollstock for easy removal
  • Pressure sensitive with a permanent adhesive, adheres instantly to waste bags, containers and disposables
  • Affix to containers of medical material, refrigerators and freezers containing blood, or any other potentially infectious material including containers used to store, transport or ship infectious materials

Specifications

Certifications/Compliance DOT, IATA
Adhesive Type Self-adhesive
Format Label
Product Type Biohazardous Material Label
For Use With (Application) Used for affix to containers of medical material, refrigerators and freezers containing blood or any other potentially infectious material including containers used to store, transport or ship infectious materials
Quantity 1000/Roll
Size (English) 3 x 2 in.
Size (Metric) 7.62 x 5.08 cm
Length (English) 3 in.
Width (English) 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.