Learn More
Therapak™ Car Caddy Systems
Large-capacity specimen transport containers designed to fit most courier vehicles
Supplier:  Therapak™ 31134
 View more versions of this product
Catalog No. 22-131-413
Only null left
Add to Cart
Description
Description
  • Offer multiple temperature zones within the same container
  • Specimens requiring freezing, refrigerated, and ambient temperatures can be easily segregated; required temperatures of the individual specimen can be maintained
  • Interlocking foam lids help to preserve temperature; nylon mesh belt with a quick-release buckle fits across lid for added security
  • Durable, easy-to-clean corrugated plastic outer shell
  • Built-in reinforced metal handles assure safe transport from the vehicle to the testing area
  • Available with EPS foam or high R-value urethane inserts
Order Info
Measure 28L x 19W x 14 in. D.
Specifications
Specifications
1/3 urethane insert
Large capacity courier transports
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.