Learn More
Therapak™ Kidney Stone Collection Cup
Catalog No. 22131485 Shop All Therapak Corporation Products
Change view
Click to view available options
No. per Pack:
12/Pk.
Catalog No. No. per Pack
22-131-485 12/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-131-485 Supplier Therapak™ Supplier No. 75109
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Nylon screen at the bottom of the cup for stone capture
For use with patients attempting to pass stones following extracorporeal shock wave lithotripsy
The StoneScreen™ kidney stone collection cup was constructed to capture renal calculi.
  • Collapsible plastic strainer cup
  • Designed for both men and women
  • Once the stone is captured, it can be placed in the storage space under the lid and saved for analysis
Specifications
Description Kidney Stone Collection Cup
For Use With (Application) Capture renal calculi from patients attempting to pass stones following extracorporeal shock wave lithotripsy
Includes Collapsible plastic strainer cup with nylon screen at the bottom of the cup for stone capture and storage space under the lid
No. per Pack 12/Pk.