Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Double-Up Arrow Marking Label

Catalog No. 22130120 Shop All Therapak Corporation Products
Change view
Click to view available options
Size (Metric):
7.62 x 10.1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric)
22-130-120 7.62 x 10.1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-130-120 Supplier Therapak™ Supplier No. 54635
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Part of IATA 650 packing instructions for specimen transport

  • Marking labels guide upright orientation of shipping boxes
  • Apply to opposite ends of corrugated box
  • Pressure sensitive with a permanent adhesive
  • In accordance with IATA, proper labeling for compliant specimen shipping systems includes application of these orientation labels to opposite ends of outer packaging

Specifications

Certifications/Compliance DOT, IATA
Adhesive Type Self-adhesive
Format Label
Product Type Double Arrow Up Orientation Label
For Use With (Application) Used to transport biological specimens
Quantity 500/Pk.
Size (English) 3 x 4 in.
Size (Metric) 7.62 x 10.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.