Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Duramark™ Fabric Lock Boxes

Catalog No. 22131430 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
Lock Box
Lock Box, Zipper Lock, Keys
Lock Box, Zipper Lock, Keys, Strap
Size:
Small
Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
22-131-430 Lock Box, Zipper Lock, Keys, Strap Large
22-131-425 Lock Box Small
22-131-426 Lock Box, Zipper Lock, Keys Small
22-131-427 Lock Box, Zipper Lock, Keys, Strap Small
22-131-428 Lock Box Large
22-131-429 Lock Box, Zipper Lock, Keys Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-131-430 Supplier Therapak™ Supplier No. 36505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Available in standard small and large sizes

  • Soft sides insulated with 1/4 in. foam
  • Consists of zipper closure, mesh envelope to contain refrigerant packs, and an adjustable strap for ease of courier handling during pickup activity
  • Optional zipper lock includes two matching keys, and optional security strap, which is reinforced with a steel cord sewn into the strap, helps assure the sample will be safe and secure
Standard color: Royal Blue Regency
Standard printing: Silver text with red biohazard symbol on one side only
Options include: Zipper lock, security strap
Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features (i.e. number of pockets), closure (i.e., Zipper, Velcro, buckle, etc.) and print
Minimum customization order quantity is 24 bags.
Artwork creation is available.
Small Size Dimensions: 9 x 8 x 7 in. (22.86 x 20.3 x 17.7cm)
Large Size Dimensions: 11 x 9 x 7 in. (27.9 x 22.86 x 17.7cm)

Specifications

Includes Lock Box, Zipper Lock, Keys, Strap
Color Royal Blue Regency
Size Large
Type With Red Biohazard Symbol
Dimensions (LxWxH) 11 x 9 x 7 in.
Material Fabric
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.