Learn More
Description
Includes
8 oz. cold packs, specimen bag, Absorbent material, Exempt Animal Specimen label, Biohazard label, Cushioning material, Packing instructions
The International Air Transport Association (IATA) Dangerous Goods Regulations (DGR) manual contains a category that was created for shipping animal specimens for which there is minimal likelihood that pathogens are present. The regulations cite examples of many routine laboratory tests that can now be transported as Exempt Animal Specimens (EAS). Examples of those tests are: blood or urine for cholesterol levels, hormone levels, prostate specific antigens (PSA), tests to monitor organ function (heart, liver, kidney), tests conducted for pregnancy and biopsies for cancer.
Standard small insulated cooler and box
Meets following IATA Exempt Animal Specimen package requirements:
- 100 × 100mm dimension on one side
- Leakproof primary container
- Leakproof secondary container
- Absorbent between primary and secondary sufficient to absorb total primary container(s) contents
- Outer packaging of adequate strength for its capacity
Order Info
Cooler L x W x H: 11 x 9 x 11 in. (exterior), 8 x 6 x 8 in. (interior)
Specifications
Specifications
| Includes | 8-oz cold packs, specimen bag, absorbent/cushioning material, Exempt Animal Specimen and biohazard labels, packing instr. |
| Type | Refrigerated Shipper |
| Dimensions (LxWxH) | 8 x 6 x 8 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.