Learn More
Therapak™ Exempt Animal Specimen (EAS) Ambient Shipping Systems
Catalog No. 22130436 Shop All Therapak Corporation Products
Change view
Click to view available options
Quantity:
Up to six tubes
Catalog No. Quantity
22-130-436 Up to six tubes
1 options
This item is not returnable. View return policy
Catalog No. 22-130-436 Supplier Therapak™ Supplier No. 56882/DUP
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in both regional and national veterinary testing laboratories and hospitals
Used to ship animal specimens that don't require refrigeration
  • Small Tube Shipping System features an Aqui-Pak™ segmented absorbent pouch that holds up to four standard blood or urine tubes. Includes pre-assembled boxes, specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label, and cushioning material.
  • Medium Tube Shipping System features an Aqui-Pak segmented absorbent pouch that holds up to six standard blood or urine tubes. Includes pre-assembled boxes, specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label, and cushioning material.
  • Small Liquid Specimen Shipping System is designed to ship one standard or larger (30 to 60mL) liquid specimen container. Includes pre-assembled boxes, specimen transport bag, absorbent material, Exempt Animal Specimen label, biohazard symbol label, and cushioning material.
  • Large Multisample Shipping System accommodates multiple specimen transport bags for consolidation at collection site and shipping to testing laboratory. Includes a large transport box with easy lock flaps, a 13 × 18 in. biohazard transport bag, 12 × 12 in. absorbent sheet, Exempt Animal Specimen label, and biohazard symbol label. Holds up to ten 6 × 9 in. specimen transport bags.
  • TearTech™ Specimen Shipping System features a bulk-pack case of shipping supplies to transport ambient samples. Each TearTech box holds up to four specimen bags. Includes 50 TearTech boxes, 100 zip-style biohazard transport bags (two-pocket bags to hold sample and requisition form), 100 Aqui-Pak segmented absorbent pouches (each Aqui-Pak holds up to 6 standard blood or urine tubes), 50 biohazard labels, 50 Exempt Animal Specimen labels, and cushioning material.
Compliance
All systems comply with IATA regulations for ground and air shipments of specimens.
Specifications
Quantity Up to six tubes
Applications Blood or urine tubes
Type Medium Ambient IATA Transport Box
Dimensions (LxWxH) 7 x 4 x 4 in.
Capacity 6 tubes
Material Cardboard