Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Foam Tube Mailers

Catalog No. 22131477 Shop All Therapak Corporation Products
Change view
Click to view available options
No. per Case:
50/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
22-131-477 50/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-131-477 Supplier Therapak™ Supplier No. 44903
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects fragile specimens and evacuated blood tubes on outbound and inbound shipments

  • Constructed of EPS
  • Holds three tubes
  • Foam lid sits tightly over tubes to provide an insulated transport environment
  • Holds tube sizes up to 16 x 100mm

Specifications

Material EPS
Description Small foam mailer
For Use With (Equipment) Tubes up to 16 x 100 mm
Holds Up to 3 tubes
No. per Case 50/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.