Learn More
Therapak™ Foil-Top Collection Kits
Catalog No. 22130409 Shop All Therapak Corporation Products
Change view
Click to view available options
Product Type:
Single Foil-top Collection Kit
Split Foil-top Collection Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-130-409 Single Foil-top Collection Kit
22-130-410 Split Foil-top Collection Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-130-409 Supplier Therapak™ Supplier No. 39448
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
All kits include one 8 oz. collection cup with pour spout and temperature strip; one 45mL tamper-evident, flip-top transport vial graduated at 15 and 30mL; and a transport bag with absorbent material. Split-specimen kit, designed for testing that requires two samples from a single donor, includes two transport vials.
Available in single- or split-specimen collection kits
  • For drugs of abuse testing applications
  • Kit contents conveniently sealed inside the collection cup with peel-off foil lid
Specifications
Product Type Single Foil-top Collection Kit
For Use With (Application) For drugs of abuse testing
Includes 8 oz. Collection cup with pour spout, temperature strip, one 45 mL tamper-evident, flip-top transport vials, transport bag with absorbent