Learn More
Therapak™ Shipper, Side by Side Combo
Catalog No. 22130416 Shop All Therapak Corporation Products
Change view
Click to view available options
Type:
Shipper, Side by Side Combo
Catalog No. Type
22-130-416 Shipper, Side by Side Combo
1 options
This item is not returnable. View return policy
Catalog No. 22-130-416 Supplier Therapak™ Supplier No. 56554
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
  • Each box has a specific tab-lock mechanism to keep boxes closed during transport
  • Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
  • Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Specifications
Applications For diagnostic specimen shipments
Type Shipper, Side by Side Combo
Dimensions (LxWxH) 27.75 x 26 x 15.5 in.