Description
- Preprinted list allows user to quickly check appropriate items for each shipment
- Card should be placed between secondary and outer packaging
Compliance
Meets IATA Packing Instruction 650 requirement for an itemized list of contents.
Specifications
Specifications
| Certifications/Compliance | Meets IATA Packing Instruction 650 requirement for an itemized list of contents |
| Applications | For inclusion with Biological Substances shipments by air |
| Type | Shipment Contents Card |
| Material | Cardboard |