Learn More
Therapak™ Combination Temperature Biological Substance Category B Shipping Systems
Catalog No. 22130033 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
2 Med 95kPa Canister; 2 Aqui-Pak
Med 95kPa Canister, Bag; 2 Aqui-Pak
Conditions:
Frozen/Ambient
Frozen/Refigerated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Conditions
22-130-033 2 Med 95kPa Canister; 2 Aqui-Pak Frozen/Refigerated
22-130-034 2 Med 95kPa Canister; 2 Aqui-Pak Frozen/Ambient
22-130-424 Med 95kPa Canister, Bag; 2 Aqui-Pak Frozen/Ambient
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-130-033 Supplier Therapak™ Supplier No. 37810
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Transport box; UN3373 Biological Substance, dry ice label, and orientation arrow labels; 95kPa canisters/bags; absorbent material; list of contents card; cushioning material; and packing instructions
Certified for ground and air shipments of Biological Substance Category B specimens
  • DOT and IATA Packing Instruction 650 compliant for the shipment of frozen/refrigerated or frozen/ambient specimens in one package
  • Suitable for overnight shipments
Specifications
Quantity 1 Ea.
Includes 2 Med 95kPa Canister; 2 Aqui-Pak
Content And Storage Two medium 95kPa canister; 2 Aqui-Pak absorbents, cushioning; biohazard, Biological Substance Category B, arrow labels, list contents card, transport box
Capacity 12 Tubes (6 per side)
Conditions Frozen/Refigerated