Learn More
Therapak™ Refrigerated Biological Substance Category B Shipping Systems
Catalog No. 22130421 Shop All Therapak Corporation Products
Change view
Click to view available options
Capacity:
Holds 18 Tubes
Holds 6 Tubes
Includes:
3- 95 kPa Bags, 3- Aqui-Pak pouches
95 kPA Canister, Aqui-Pak pouch
95 kPa Bag, Aqui-Pak pouch
95 kPa Canister, 3 Aqui-Pak pouch
Packaging:
Corrugated box
Transport box
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes Packaging
22-130-421 Holds 18 Tubes 3- 95 kPa Bags, 3- Aqui-Pak pouches Transport box
22-130-028 Holds 6 Tubes 95 kPA Canister, Aqui-Pak pouch Transport box
22-130-032 Holds 18 Tubes 95 kPa Canister, 3 Aqui-Pak pouch Transport box
22-130-400 Holds 6 Tubes 95 kPa Bag, Aqui-Pak pouch Transport box
22-130-116 Holds 6 Tubes 95 kPa Bag, Aqui-Pak pouch Corrugated box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-130-421 Supplier Therapak™ Supplier No. 37816
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
All systems include transport box with UN3373 Biological Substance, biohazard and orientation arrow labels; EPS foam cooler, gel refrigerant pack(s), 95kPa bags or canisters, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), absorbent material, list of contents card, cushioning material, and packing instructions.
Certified for ground and air shipments of Biological Substance, Category B specimens.
  • DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
  • Systems suitable for overnight shipments; Mfr. No. 38028 field-tested for 2-8°C temperature maintenance for 48 hours of transport
Specifications
Quantity 1 Ea.
Includes 3- 95 kPa Bags, 3- Aqui-Pak pouches
Certifications/Compliance DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
Type Refrigerated Shipping for Biological Substance Category B
Content And Storage Transport box (OD: 15 x 13 x 12 in.), medium cooler (ID: 12 x 10 x 9 in.), three 95kPa bags, three Aqui-Pak™ segmented absorbent pouches, 8 oz. refrigerant packs, cushioning material, list of contents card and labels for IATA PI650 marking and compliance. 
Capacity Holds 18 Tubes
Packaging Transport box