Learn More
Therapak™ Upright Large Shipping Boxes
Catalog No. 22131490 Shop All Therapak Corporation Products
Change view
Click to view available options
Type:
Transport Box for Large Canister
Upright Large Shipping Boxes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-131-490 Upright Large Shipping Boxes
22-131-408 Transport Box for Large Canister
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-131-490 Supplier Therapak™ Supplier No. 23963G
This item is not returnable. View return policy
Each box has a specific tab-lock mechanism to keep boxes closed during transport. These ambient shipping boxes are ideal for kit packaging and outbound shipping of diagnostic kits. Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping.
Specifications
Size Large
Type Upright Large Shipping Boxes
Dimensions (L x W x H) Exterior 5.3 x 5.3 x 7.375 in.
Dimensions (LxWxH) Inner: 5.44 x 5.44 x 7.5 in.