Learn More
Therapak™ Urinalysis (UA) Transport System
Catalog No. 22130408 Shop All Therapak Corporation Products
Change view
Click to view available options
Volume (Metric):
12 mL
Catalog No. Volume (Metric)
22-130-408 12 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-130-408 Supplier Therapak™ Supplier No. 94500
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Ideal for use in clinical trial site and testing laboratory
  • Maintain viability of urine specimens for max. of 96 hr. without refrigeration
  • Involves three steps
    • Step 1: fill beaker collection cup with urine
    • Step 2: trasfer urine fro cup to transport tube
    • Step 3: replace the cap of the tube untill both surfaces meet
    Specifications
    Product Type Urinalysis Transport System
    For Use With (Application) For use in clinical trial site and testing laboratory
    Includes Conical style tube, push cap with spout, instruction label and preservative tablet
    Volume (English) 0.03 gal.
    Volume (Metric) 12 mL