Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Zip Bag with Biohazard Symbol

Catalog No. 22130111 Shop All Therapak Corporation Products
Change view
Click to view available options
Dimensions (L x W):
13 x 18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-130-111 13 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-130-111 Supplier Therapak™ Supplier No. 15522
Add to Cart
Add to Cart
This item is not returnable. View return policy

For large capacity shipments of multiple sample packages from a single site or multiple sites to a testing laboratory

Specifications

Autoclavable Not Autoclavable
Color Clear
For Use With (Application) Used for large capacity shipments of multiple sample packages from a single site or multiple sites to a testing laboratory
Includes Biohazard symbol, zip top, 2 pockets
Material LDPE
Dimensions (L x W) 13 x 18 in.
Product Type Biohazard Transport Bag
Length (Metric) 328 mm
Width (Metric) 485 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.