Learn More
Thermco Products Inc Melt Point 0/160C x 1.0C (Encompass)
Supplier:  Thermco Products Inc ACCFISJOH1
Encompass_Preferred
Thermometer manufactured to be used in a Fisher-Johns Melting Point Apparatus. Fiher catalog number 12-144 and 12-144-1. made from the finest thermometric tubing and has been enealed. Thermometer is eligible for N.I.S.T. certification
Catalog No. NC0521237
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.