Learn More
Stoelting™ Thermal Cautery Instrument
SDP
Stoelting™ Thermal Cautery Instrument is line-powered cautery and maintains the desired temperature, from 300 to 2200°F consistently during tissue contact and does not rapidly diminish.
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-001-018
SDP
View Documents
Stoelting™
59014
Reusable tips N/A N/A N/A
Description
Description
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.