Learn More
GA International THERMAL CRYO LABELS1000L WH
Supplier:  GA International DTU85NOTC11WH
Encompass_Preferred
CryoSTUCK® - Direct Thermal Cryo Labels for Frozen Vials and Surfaces, 2" X 0.625" /50.8 mm X 15.9 mm White, 1" Core, Notch, 1000 L/R
Catalog No. NC2885846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.