Learn More
Andwin Scientific THERMAL CRYOTAG 1.5X0.5 750PK
SDP
Supplier:  Andwin Scientific L10072
Encompass_Preferred
The 1.5X0.5 in. White Direct Thermal Cryotag can withstand temperature ranging from -196- 70 deg.C. It adheres most surfaces without cracking, peeling, degrading, scuffing or smearing. It is sold in a pack of 750.
Catalog No. 50-999-403
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.