Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Thermal Linear Expansion Replacement Rods
SCI_ED

Catalog No. p-7203236 Shop All Eisco Products
Change view
Click to view available options
Material:
Aluminum
Brass
Copper
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S27633 Aluminum
S27634 Brass
S27635 Copper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S27633 Supplier Eisco™ Supplier No. LEAL
Only null left
Add to Cart
Add to Cart

Thermal Linear Expansion Replacement Rods to be used with Eisco™ Labs Linear Expansion Apparatus (S23742 and S96188)

  • For use with Eisco™ Linear Expansion Apparatus (S23742 and S96188)
  • Dimensions: 9.7 x 0.25 in.
  • Sold individually

Specifications

Grade 10 to Undergraduate
Age 14 to 20
For Use With (Equipment) Eisco™ Linear Expansion Apparatus (S23742 and S96188)
Includes 1 Rod
Material Aluminum
Weight (English) 0.8 lb.
Weight (Metric) 362.87 g
Class Size 25 Students
Product Type Thermal Linear Expansion Replacement Rods
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.