Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience Thermal Transfer Film
SDPSCI_ED

Catalog No. 16949099 Shop All Interscience Products
Change view
Click to view available options
For Use With (Equipment):
DataLink Pro
Thermal Printer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-949-099 Thermal Printer
16-949-100 DataLink Pro
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-949-099 Supplier Interscience Supplier No. 410130
Add to Cart
Add to Cart

Thermal transfer printer ribbon for thermal printers.

  • Thermal transfer printer ribbon
  • Available for Interscience thermal printers or DataLink Pro

Specifications

Product Type Thermal Transfer Printer Ribbon
Color Black
For Use With (Equipment) Thermal Printer
Print Technology Thermal Transfer
Autoclavable Not Autoclavable
Sterility Non-sterile
Format Roll
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.