Learn More
Thermco Products Inc Rapid response blood bank immunohematology thermometer. 0 deg./40 deg.C, 1.0C div, +/-1 degree accuracy. Place bags on the insulated pad and receive an accurate temp reading within 10 sec.
Supplier:  Thermco Products Inc HCBB1000
Encompass_Preferred
This product was recently added by customer request, and is available for your convenience. We strive to provide our customers with a one-stop shop for the entire scientific supplies category. More relevant content may be added as customer demand increases.
Catalog No. NC0559829
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.