Learn More
Thermco Products Inc NIST compliant, factory certified glass Hg thermometer, 30 deg./124 deg.F, 0.2F div, 76mm imm, certified at 32 deg.F, 40 deg.F, 60 deg.F, 100 deg.F, 120 deg.F, 460mm
Supplier: Thermco Products Inc ACC10023FC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.