Learn More
SI Analytics™ Thermo Printer and Accessories
Catalog No. p-7202952 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
Thermal Paper
Thermal Printer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
15-177-889 Thermal Printer
15-177-891 Thermal Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-177-889 Supplier SI Analytics™ Supplier No. 285220480
Only null left
Add to Cart
Add to Cart
For use with Autotitrators and T 500 Piston Burette
TRUSTED_SUSTAINABILITY
Specifications
Connector Types USB
Description Thermo Printer and Accessories
Type Thermal Printer
Length (Metric) 112 mm
Model TZ 3863
For Use With (Equipment) TitroLine™ 5000/6000/7000/7500 KF/7500 KF Trace/7750/7800, TITRONIC™ 300/500