Learn More
Thermo Scientific™ ABgene™ Strep Thermo-Fast™ PCR Plates
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Natural |
| Material | Polypropylene |
| No. of Wells | 96 |
| Surface Treatment | Streptavidin |
| Well Shape | Round Bottom (U-well) |