Learn More
Description
- Patented hot-start enzyme Thermo-Start™ DNA Polymerase provides superior specificity and sensitivity
- Buffer provides for maximal performance in singleplex and multiplex reactions
- All mixes undergo extensive QC to ensure batch-to-batch and assay-to-assay consistency
- Licensed for PCR
Specifications
Specifications
| No. of Reactions | 500 |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.