Learn More
Thermo Scientific™ ABsolute™ MAX QRT-PCR Mixes
Catalog No. AB1349B Shop All Thermo Scientific Products
Change view
Click to view available options
Capillary dUTP Mix for the Lightcycler; 16 x 1.25mL vials; 2000 reactions
Capillary dUTP Mix for the Lightcycler; 2 x 1.25mL vials; 250 reactions
QRT-PCR ROX mix; 1.2mL vials; 800 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AB-1349B Capillary dUTP Mix for the Lightcycler; 16 x 1.25mL vials; 2000 reactions
AB-1340B QRT-PCR ROX mix; 1.2mL vials; 800 reactions
AB-1349A Capillary dUTP Mix for the Lightcycler; 2 x 1.25mL vials; 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AB-1349B Supplier Thermo Scientific™ Supplier No. AB1349B
Provide a quick, reliable, and efficient method for detecting and quantifying RNA
  • Formulated for use in one-step QRT-PCR reactions, with gene-specific primers and probes
  • Minimal pipetting required—protocol is easy to follow and automation is viable
  • 2X mix is supplied with a proprietary blend of RTases combining the advantages of AMV and MMuLV RTases to give unparalleled performance
  • Can be used at elevated temperatures, avoiding problems caused by secondary structure in the RNA template
  • QRTase Blend has greater dynamic range than either AMV or MMuLV, leading to greater accuracy in QRT-PCR experiments
TRUSTED_SUSTAINABILITY
Specifications
No. of Reactions 2000
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.