Learn More
Thermo Scientific™ ABsolute™ QPCR Low ROX™ Mixes
Catalog No. AB1320A
Change view
Click to view available options
:
dUTP, 100 reactions; 2 x 1.25mL
dUTP, 200 reactions; 1 x 5mL
dUTP, 2000 reactions; 10 x 5mL
dUTP, 800 reactions; 16 x 1.25mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AB-1320-A dUTP, 100 reactions; 2 x 1.25mL
AB-1320-B dUTP, 800 reactions; 16 x 1.25mL
AB-1321-A dUTP, 200 reactions; 1 x 5mL
AB-1321-B dUTP, 2000 reactions; 10 x 5mL
AB-1324-A dUTP, 100 reactions; 2 x 1.25mL
AB-1324-B dUTP, 800 reactions; 16 x 1.25mL
AB-1325-A dUTP, 200 reactions; 1 x 5mL
AB-1325-B dUTP, 800 reactions; 16 x 1.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. AB-1320-A Supplier Thermo Scientific™ Supplier No. AB1320A
Optimized for use with real-time instruments that are able to detect long wavelength dyes with greater sensitivity (ABI™ 7500)
  • Incorporate Thermo-Start™ DNA polymerase, a hot-start enzyme that provides superior specificity and excellent sensitivity
  • Utilize a proprietary reaction buffer that provides high sensitivity, specificity and reproducibility
  • Contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes
  1. Practice of the patented 5apostrophe Nuclease Process requires a license from Applied Biosystems. The purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaserapostrophes own internal research when used with the separate purchase of Licensed Probe. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained from the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
  2. Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaserapostrophes own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
TRUSTED_SUSTAINABILITY
Specifications
No. of Reactions 100