Learn More
Thermo Scientific™ AMS-4 Continuous Beta Air Monitors
Description
Continuous air monitor (CAM) offering beta particulate, radioiodine, or noble gas detection.
- Detachable sampling head
- Reports both DAC and cpm
- Real time gamma background subtraction
- Stand alone or network configuration
- Mass flow measurements
- Fast and slow concentration alarms, cpm alarm, DAC-hour alarm
- Computer calibration
- Can also be used for stack monitoring of effluent releases using the in-line detector head
- Keypad: 18 keys, full numeric plus function keys
- Status Indicators: Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
- Printer: RS-232 serial DB9
- Data Log Buffer: 2000 entries of combined Concentration and Status
- Change information: Equivalent to 83 days of 1 hour detail, nearly 14 days of 10 mintes detail or over 24 hours of 1 minute detail
- Alarm/Fail Relays: SPST, 15A at 30VDC/120VAC resistive
- Analog Output: 0 to 5VDC output, five decade logarithmic
Compliance
Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
Specifications
Specifications
| Product Type | In-line Entry Sampling Head |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.