Learn More
Thermo Scientific™ ART™ Non-Filtered Pipet Tips with MicroPoint™ Tip Design
GREENER_CHOICE
Catalog No. 2123677C
Change view
Click to view available options
Volume (Metric):
5 to 200 μL
Catalog No. Volume (Metric)
21-236-77C 5 to 200 μL
1 options
Catalog No. 21-236-77C Supplier Thermo Scientific™ Supplier No. 3542
Add to Cart
Add to Cart
MicroPoint design means the tip profile is nearly 50% smaller than standard blunt-end tips and significantly more narrow than beveled-end tips
  • Same superior quality and performance as all Thermo Scientific ART pipet tips
  • Crystal clear tips allow you to easily see transparent samples
  • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting the desired volume of liquid
  • ART 10 tips are ideal for low volume samples
  • The extended length ART 10 REACH™ and ART 1000 REACH prevent contamination of samples, tubes, and pipetters
  • Ultra Micro and Pure Ultra Micro are also well suited for work with low-volume samples
  • ART 200 are equivalent to the “Universal Yellow“ tip and have a reference mark at 10μL
ART Non-Filtered tips are available in yellow and blue colors, graduated, beveled tips and large volumes..
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 5 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color White
Autoclavable Autoclavable
Model XLP
Format Rack
Tip Style Extended Length
Product Line ART
Material Polypropylene