Learn More
Thermo Scientific™ ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips

Description
Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.
- Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
- Ideal for genomic work
- Universal Tip, fits most brands of pipettes
- ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
- Non-filtered tip
- ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
- Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
Specifications
| Description | ART 1000G, Non-filtered, Sterile, Racked, Lift-off Lid |
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Model | ART 1000G |
| Clean Claims | DNase, RNase, DNA, ATP and Endotoxin Free |
| For Use With (Application) | Genomic workflows, fragile cell lines, viscous liquids |
| Features | Wide bore orifice |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.