Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Non-Filtered, Wide-Bore, Genomic Pipette Tips
GREENER_CHOICE

Catalog No. 2123678C
Change view
Click to view available options
Volume (Metric):
200 μL
1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Lift-off Lid Rack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
21-236-78C 1000 μL Non-sterile Lift-off Lid Rack
21-236-76B 200 μL Sterile Lift-off Lid Rack
21-236-78A 1000 μL Non-sterile Bulk
21-236-78B 1000 μL Sterile Lift-off Lid Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-236-78C Supplier Thermo Scientific™ Supplier No. 3592
Add to Cart
Add to Cart

Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows.

Thermo Scientific™ ART™ Non-filtered, wide bore pipette tips are ideal for pipetting in genomic workflows. The wide orifice prevents shearing of cells and DNA and RNA creating longer, continuous strands. The wide opening also allows for easier pipetting of viscous fluids or solutions. Ideal for use in reference labs and blood banking applications.

  • Wide orifice is larger than standard tip openings making it ideal for pipetting fragile cell lines and viscous samples
  • Ideal for genomic work
  • Universal Tip, fits most brands of pipettes
  • ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
  • Non-filtered tip
  • ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
  • Lift-off lid racks provide access to all tips, great for multichannel pipetting
TRUSTED_SUSTAINABILITY

Specifications

Description ART 1000G, Non-filtered, non-sterile, Racked, Lift-off Lid
Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model ART 1000G
Clean Claims DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
For Use With (Application) Genomic workflows, fragile cell lines, viscous liquids
Features Wide bore orifice
Format Lift-off Lid Rack
Tip Style Wide Orifice
Packaging 3200/Pk.
Quantity 3200/Pk.
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.