Learn More
Thermo Scientific™ ART™ Solvent Safe Carbon Filtered Pipette Tips
Description
For acids, bases and aggressive organic solvents
Revolutionary filter material handles the pipetting rigors of Combinatorial Chemistry. Unique use of the ART™ filter with Folded Activated Carbon offers both pipetter and sample protection against destructive carryover aerosols and vapors from volatile organic solvents, all without compromising pipetting accuracy and precision.
- For manual pipetting of organic solvents
- Utilizes new formula of the ART (aerosol resistant tip) filter tip
- Helps to eliminate pipetting failures and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
- Proprietary blend of Folded Activated Carbon with a 40-micron polyethylene filter provides effective solvent protection
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| Filtered | Filtered |
| Sterility | Non-sterile |
| Format | Bulk |
| Tip Style | Universal Fit |
| For Use With (Equipment) | Universal Fit |
| Product Line | ART |
| Material | Carbon with Polyethylene Filter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.