Learn More
Thermo Scientific™ Barnstead Nanopure Consumables Packages
Description
Includes
Cartridge for R/O or distilled water feed (D50280), UV replacement lamp (LMX13), and sanitization kit (CMX25). Life Science packages add replacement ultrafilter membrane (FL1192X1).
For use with Nanopure systems. Choose from UV or Life Science packages for R/O distilled or deionized feeds.
Nanopure UV Package for R/O Distilled Feed
- Includes Durapack™ organic-free R/O cartridge pack, UV replacement lamp and sanitization kit
- Save 13%
- Includes Durapack™ organic-free deionization cartridge pack, UV replacement lamp and sanitization kit
- Save 12%
- Includes Durapack™ organic-free R/O cartridge pack, UV replacement lamp, sanitization kit and replacement ultrafilter membrane
- Save 15%
- Includes Durapack™ organic-free deionization cartridge pack, UV replacement lamp, sanitization kit and replacement ultrafilter membrane
- Save 15%
Specifications
Specifications
| For Use With (Equipment) | Nanopure Systems, R/O Distilled |
| Includes | Durapack™ Organicfree R/O cartridge pack, UV replacement lamp and sanitization kit |
| Type | Nanopure UV Package |