Learn More
Thermo Scientific™ BetaMax Drop-In Acid Guard Cartridges
Description
Requires
Thermo Scientific Uniguard direct-connection or stand-alone holder, sold separately.
- The 10mm design offers maximum protection with minimal increase in retention
- Fits Thermo Scientific Uniguard direct-connection and stand-alone holders
Features of BetaMax Acid Solid Phase
- Designed for acidic analytes
- Ultra-high surface coverage for maximum retention
- Excellent stability for long column lifetimes
Specifications
Specifications
| Particle Size | 5 μm |
| Length (Metric) | 10 mm |
| Diameter (Metric) Inner | 2.1 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.