Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ CN Drop-in Guard Cartridges

Catalog No. 03051747 Shop All Thermo Scientific Products
Change view
Click to view available options
3.0 x 10mm ID x L; 5μm particle size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
03-051-747 3.0 x 10mm ID x L; 5μm particle size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-051-747 Supplier Thermo Scientific™ Supplier No. 73405014001

missing translation for 'validMainframeMsg'

Requires

Thermo Scientific Unigard™ direct connection or stand-alone holder

Offer convenience, economy and effective protection for extending column lifetime

  • The 10mm design offers maximum protection with minimal increase in retention
  • Compatible with Thermo Scientific Uniguard™ direct connection and stand-alone holders

Features of BioBasc CN Solid Phase

  • 300Å pore size for improved biomolecule separations
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Elution order changes
  • Outstanding reproducibility, efficiency and column lifetime

Order Info

Requires the use of the Thermo Scientific UNIGUARD™ direct connection or stand alone holder.

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 3 mm
Stationary Phase CN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.