Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ Phenyl Drop-in Guard Cartridges

Catalog No. p-3647694
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
2.1 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
03-051-723 1 mm
03-051-724 2.1 mm
03-051-726 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-051-723 Supplier Thermo Scientific™ Supplier No. 72405011001

Requires

Thermo Scientific UNIGUARD™ direct connection or stand alone holder

Offer convenience, economy and effective protection for extending column lifetime

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
  • Available in packs of 4

Features of BioBasic Phenyl Solid Phase

  • 300Å pore size for maximum performance with biomolecules
  • Lower carbon load for optimal retention of larger peptides and proteins
  • Offers different selectivity to alkyl chain chemistires for critical separations
  • Excellent for LC/MS separations

Order Info

Thermo Scientific UNIGUARD™ direct connection or stand alone holder

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 1 mm
Stationary Phase Phenyl
Product Line BioBasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.