Learn More
Epredia™ Permafrost™ Slides
Catalog No. 4441 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Gold
Green
Lavender
Orange
Pink
Red
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
44-41 Aqua
44-44 Lavender
44-45 White
44-47 Yellow
44-50 Green
44-40 Tan
44-42 Gold
44-43 Red
44-48 Orange
44-46 Blue
44-51 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 44-41 Supplier Epredia™ Supplier No. 4441
Add to Cart
Add to Cart
This item is not returnable. View return policy
Available in a wide variety of colors to ease identification.
  • Resistant to commonly used laboratory solvents
  • 1.0 to 1.2 mm thick with ground edges
Specifications
Frosted Permafrost 1 End
Material Glass
Thickness (Metric) 1 to 1.2 mm
Edges Ground
Color Aqua
Quantity 1440/Cs.