Learn More
Thermo Scientific™ CryoMed™ Thermocouple Sensors
Catalog No. TF4000381
Change view
Click to view available options
Type:
Ribbon Type
Thermocouple Sensor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
TF4000381 Thermocouple Sensor
TF4000393 Ribbon Type
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TF4000381 Supplier Thermo Scientific™ Supplier No. 4000381
Only null left
Add to Cart
Add to Cart
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
  • Include male plug
For controlled-rate freezing applications
TRUSTED_SUSTAINABILITY
Specifications
Type Thermocouple Sensor
For Use With (Equipment) CryoMed 7450