Learn More
Thermo Scientific™ DELTABOND Resolution AK Drop-In Guard Cartridges
Description
Requires
Thermo Scientific UNIGUARD™ direct connection or standalone holder, sold separately.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD direct connection and standalone holders
- Columns dedicated to aldehyde and ketone analysis
- Ideal for auto emissions separations
- For high throughput analysis of simple mixtures
Specifications
Specifications
| Particle Size | 5 μm |
| Length (Metric) | 20 mm |
| Diameter (Metric) Inner | 4.6 mm |
| Stationary Phase | Resolution AK |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.