Learn More
Thermo Scientific™ Disposable Inoculator Assembly for 96-Well Plates
Shop All Thermo Scientific Products
Description
For bacterial studies, blood toxicity and many tissue culture applications
Simultaneously transfers bacterial colonies from a vessel of choice to a 96-well Microtiter™ format.
- Sterilized by gamma radiation
- For use with inoculum tray
- Includes 95 tips and one control/blank
- 10μL/tip
Designed for bacterial studies, blood toxicity and many tissue culture applications.
Order Info
Case of 50
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Sterility | Sterile |