Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Easy Peel Heat Sealing Foil

Catalog No. AB0745 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB-0745 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB-0745 Supplier Thermo Scientific™ Supplier No. AB0745
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for long-term, low-temperature storage.

Suitable for long-term, low-temperature storage.

  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20μm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
TRUSTED_SUSTAINABILITY

Specifications

Color Foil
Solvent Or Alcohol Resistant Life Science Reagents Resistant, DMSO Resistant
Material Aluminum
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
Dimensions (L x W) 85 x 135 mm
Quantity 100 Pk.
For Use With (Application) Ideal for long term storage at low temperatures, PCR
Content And Storage General, Long Term
Format Sheets
For Use With (Equipment) ALPS 25 Automated Laboratory Plate Sealer
Product Type Easy Peel Film Laminate
Product Line Abgene
Adhesive Polypropylene
Certified RNase/DNase-free No
Pierceable No
Temperature Range (Metric) -200°C to +90°C, -200°C to +120°C with adequate pressure
Well Shape All well shapes
Plate Compatibility Polypropylene, Polyethylene, Cyclic Olefin Copolymer
Incubation Conditions General
Peelable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.