Learn More
Thermo Scientific™ Electrophoresis Power Supplies
Catalog No. FB105 Shop All Thermo Scientific Products
Change view
Click to view available options
Model 105
Catalog No.
FB-105 Model 105
1 options
Catalog No. FB-105 Supplier Thermo Scientific™ Supplier No. EC105115/PRC
Suitable for a wide range of applications
  • Designed for safety, with no-load and failure notification
  • Overvoltage protection and jacks that are set away from controls
Low Voltage Dual Mode Power Supply, Model 105ECA-LVD
  • Ultra-compact, low-voltage unit is easy to operate with only one switch and one adjustment knob to control the unit's voltage output
  • LED display toggles between output voltage or output current
  • Ideal for submarine gels, mini SDS-PAGE, mini tank (wet) electroblotting and pulsed-field electrophoresis (requires pulse controller)
Standard Dual Mode Power Supplies
  • Automatic power shutoff at end of run
  • Automatic recovery after power failure
  • Backlit LED display, mode indicator lights, timer and multiple output jacks
  • Soft-touch keypad controls Memory retains settings after shutdown Stackable design allows use of multiple units in less space
  • Locking kickstand permits positioning the unit at an angle for better viewing
  • Nonskid rubber feet provide stability
High-Current Models EC 200XL/200XL2
  • Utilize a numeric keypad
  • Suitable for high-current electroblotting (Western tank and semidry, Southern and Northern) and SDS-PAGE applications
High-Voltage Models EC 300XL/300XL2
  • Use UP/DOWN arrow controls
  • Suitable for SDS-PAGE, DGGE/CDGE, and DNA/RNA submarine electrophoresis
Programmable Power Supplies, Models EC 3000XL and 3000XL2
  • Additional features include a third, constant power mode, remote temperature probe and nine-program memory Feature a numeric keypad Suitable for DNA sequencing and high-voltage electrophoresis applications
Service Plan
Service agreement available.
Warranty and Services
Model 105ECA-LVD: Four-year warranty. All other models: 12-month warranty.