Learn More
Thermo Scientific™ EPD Mk2, G and N2 Readers
To complement the Thermo Scientific EP DMk2, G and N2. The basic IrDA reader, is ideal for office environments and mobile applications
Supplier:  Thermo Scientific™ EPD/1/43772/001
 View more versions of this product
Catalog No. 19-175-720
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Compatible with the full range of Mk2 software products
  • Connect directly to the PC via a serial or optional USB interface
  • Require no power supply
  • For more demanding environments, and where it is necessary to ensure that only one specific EPD can be “seen” by the reader, the desktop model ACT-5 may be used
  • The IrDA and ACT-5 are compatible with EasyEPD2 and EasyIssue software
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 in.
2.5 lb.
5 in.
Serial RS232 or USB 1.1 and 2.0 compatible interface direct to PC
For office Environments and Mobile applications
3.9 ft.
1.2 m
Act-5 Desktop Reader
7.62 cm
1.1 kg
12.7 cm
7 in.
Easy EPD software, USB cable
17.78 cm
7 x 5 x 3 in. (17.78 x 12.7 x 7.62 cm)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.